Tisk…

(S)-3-(Boc-amino)piperidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL08601-1G-FEA 0 CZK
SIAL08601-1G-F
(S)-3-(Boc-amino)piperidine, Sigma-Aldrich®
(S)-3-(Boc-amino)piperidine
Vzorec: C₁₀H₂₀N₂O₂
MW: 200,28
MDL Number: MFCD03093383
CAS číslo: 216854-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR