Tisk…

(S)-(+)-3-Amino-1-boc-piperidine, Sigma-Aldrich®

Dodavatel: Merck

SIAL19929-1G-FEA 0 CZK
SIAL19929-1G-F
(S)-(+)-3-Amino-1-boc-piperidine, Sigma-Aldrich®
(S)-(+)-3-Amino-1-boc-piperidine
Vzorec: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
MDL Number: MFCD03094718
CAS číslo: 625471-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR