1-Boc-4-(methylamino)piperidine ≥97%
H52576.03EA
1960
CZK
H52576.03
1-Boc-4-(methylamino)piperidine ≥97%
1-Boc-4-(methylamino)piperidine
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214,3046 g/mol |
MDL Number:
MFCD02259411 CAS číslo: 147539-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...