Tisk…

(S)-1-Boc-3-hydroxypiperidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL687367-1GEA 0 CZK
SIAL687367-1G
(S)-1-Boc-3-hydroxypiperidine, Sigma-Aldrich®
(S)-1-Boc-3-hydroxypiperidine
Vzorec: C₁₀H₁₉NO₃
MW: 201,27
MDL Number: MFCD04115307
CAS číslo: 143900-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR