Tisk…

4-(Bromomethyl)benzophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL449385-5GEA 5890 CZK
SIAL449385-5G
4-(Bromomethyl)benzophenone, Sigma-Aldrich®
4-(Bromomethyl)benzophenone
Vzorec: C₁₄H₁₁BrO
MW: 275,1405 g/mol
MDL Number: MFCD00092076
CAS číslo: 32752-54-8
EINECS: 251-197-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR