Tisk…

N-Phthaloylglycine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALP40506-25GEA 0 CZK
SIALP40506-25G SIALP40506-100G
N-Phthaloylglycine, Sigma-Aldrich®
N-Phthaloylglycine
Vzorec: C₁₀H₇NO₄
MW: 205,17
MDL Number: MFCD00005900
CAS číslo: 4702-13-0
EINECS: 225-177-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR