Tisk…

1-Pyrenebutanol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL399655-1GEA 3540 CZK
SIAL399655-1G
1-Pyrenebutanol, Sigma-Aldrich®
1-Pyrenebutanol
Vzorec: C₂₀H₁₈O
MW: 274,3563 g/mol
MDL Number: MFCD00192423
CAS číslo: 67000-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR