Tisk…

Tris[N,N-bis(trimethylsilyl)amide]europium(III), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL547816-1GEA 0 CZK
SIAL547816-1G
Tris[N,N-bis(trimethylsilyl)amide]europium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amide]europium(III)
Vzorec: C₁₈H₅₄EuN₃Si₆
MDL Number: MFCD03411248
CAS číslo: 35789-02-7
UN: 3396
ADR: 4.3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR