Benzophenone-4-maleimide, Sigma-Aldrich®
SIALM9775-100MGEA
0
CZK
SIALM9775-100MG
Benzophenone-4-maleimide, Sigma-Aldrich®
Benzophenone-4-maleimide
Vzorec:
C₁₇H₁₁NO₃ MW: 277,2700 g/mol |
MDL Number:
MFCD00079465 CAS číslo: 92944-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...