Tisk…

Sodium acetate-[2-¹³C] (99% ¹³C ), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL279315-1GEA 6950 CZK
SIAL279315-1G
Sodium acetate-[2-¹³C] (99% ¹³C ), Sigma-Aldrich®
Sodium acetate-[2-¹³C]
Vzorec: H₃CCOONa
Storage Temperature: Ambient
MDL Number: MFCD00037668
CAS číslo: 13291-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR