Tisk…

Adrenalone hydrochloride ≥98% (dry basis), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL02270-10GEA 0 CZK
SIAL02270-10G
Adrenalone hydrochloride ≥98% (dry basis), Sigma-Aldrich®
Adrenalone hydrochloride
Vzorec: C₉H₁₁NO₃·HCl
MW: 217,65 g/mol (anhy)
MDL Number: MFCD00035075
CAS číslo: 62-13-5
EINECS: 200-525-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR