2-Aminoazotoluene, Supelco®
Dodavatel: Merck
Synonyms:
Solvent Yellow 3
, 4′-Amino-2,3′-dimethylazobenzene
, Fast Garnet GBC base
, C.I. 11160
M-Clarity™ quality level = MQ100
31629-250MGEA
15410
CZK
31629-250MG
2-Aminoazotoluene, Supelco®
2-Aminoazotoluene
Vzorec:
C₁₄H₁₅N₃ MW: 225,29 g/mol |
MDL Number:
MFCD00007733 CAS číslo: 97-56-3 EINECS: 202-591-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...