Tisk…

Diphenylacetaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALD204250-5GEA 2840 CZK
SIALD204250-5G SIALD204250-25G
Diphenylacetaldehyde, Sigma-Aldrich®
Diphenylacetaldehyde
Vzorec: C₁₄H₁₂O
MW: 196,2400 g/mol
MDL Number: MFCD00006972
CAS číslo: 947-91-1
EINECS: 213-433-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR