Tisk…

2'-Aminobenzophenone-2-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL153273-1GEA 1070 CZK
SIAL153273-1G
2'-Aminobenzophenone-2-carboxylic acid, Sigma-Aldrich®
2'-Aminobenzophenone-2-carboxylic acid
Vzorec: C₁₄H₁₁NO₃
MW: 241,2420 g/mol
MDL Number: MFCD00007715
CAS číslo: 1147-43-9
EINECS: 214-554-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR