Tisk…

4-Allyloxy-2-hydroxybenzophenone, Sigma-Aldrich®

Dodavatel: Merck

SIAL415839-25GEA 2420 CZK
SIAL415839-25G
4-Allyloxy-2-hydroxybenzophenone, Sigma-Aldrich®
4-Allyloxy-2-hydroxybenzophenone
Vzorec: C₁₆H₁₄O₃
MW: 254,2806 g/mol
MDL Number: MFCD00191713
CAS číslo: 2549-87-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR