Tisk…

Triphenylbismuth diacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL485721-10GEA 0 CZK
SIAL485721-10G
Triphenylbismuth diacetate, Sigma-Aldrich®
Triphenylbismuth diacetate
Vzorec: C₂₂H₂₁BiO₄
MDL Number: MFCD00092697
CAS číslo: 7239-60-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR