6-Chloronicotinoyl chloride 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12354.14EA
4470
CZK
A12354.14
A12354.06
6-Chloronicotinoyl chloride 98%
6-Chloronicotinoyl chloride
Vzorec:
C₆H₃Cl₂NO MW: 176,00 g/mol Bod varu: 84…85 °C (3 mmHg) Bod tání: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051775 CAS číslo: 58757-38-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...