2-Amino-5-bromobenzoylpyridine
Dodavatel: LGC Standards PROMOCHEM
PROOMM0064.01EA
41270
CZK
PROOMM0064.01
2-Amino-5-bromobenzoylpyridine
2-Amino-5-bromobenzoylpyridine
Vzorec:
C₁₂H₉BrN₂O MW: 277,1167 g/mol |
MDL Number:
MFCD07780283 CAS číslo: 1563-56-0 EINECS: 216-352-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...