Tisk…

Geranyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL302791-5GEA 0 CZK
SIAL302791-5G
Geranyl chloride, Sigma-Aldrich®
Geranyl chloride
Vzorec: C₁₀H₁₇Cl
MW: 172,6950 g/mol
MDL Number: MFCD00000983
CAS číslo: 5389-87-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR