Tisk…

Chlorodimethyloctylsilane, Sigma-Aldrich®

Dodavatel: Merck

SIAL246859-25GEA 3930 CZK
SIAL246859-25G
Chlorodimethyloctylsilane, Sigma-Aldrich®
Chlorodimethyloctylsilane
Vzorec: C₁₀H₂₃ClSi
MW: 206,83 g/mol
MDL Number: MFCD00000511
CAS číslo: 18162-84-0
EINECS: 242-044-5
UN: 2987
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR