Isoflurane
EDQMY0000037EA
6700
CZK
EDQMY0000037
Isoflurane
Isoflurane
Vzorec:
C₃H₂ClF₅O MW: 184,49 g/mol |
MDL Number:
MFCD00066609 CAS číslo: 26675-46-7 EINECS: 247-897-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...