4′-Hydroxy-2-phenylacetophenone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIALB23077-5GEA
3750
CZK
SIALB23077-5G
4′-Hydroxy-2-phenylacetophenone, Sigma-Aldrich®
4′-Hydroxy-2-phenylacetophenone
Vzorec:
C₁₄H₁₂O₂ MW: 212,24 g/mol |
MDL Number:
MFCD00002360 CAS číslo: 2491-32-9 EINECS: 219-654-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...