Tisk…

2',4'-Dihydroxy-2-phenylacetophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL438499-1GEA 1320 CZK
SIAL438499-1G
2',4'-Dihydroxy-2-phenylacetophenone, Sigma-Aldrich®
2',4'-Dihydroxy-2-phenylacetophenone
Vzorec: C₁₄H₁₂O₃
MW: 228,2433 g/mol
MDL Number: MFCD00043754
CAS číslo: 3669-41-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR