Tisk…

1-Methoxy-2-butanol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL479756-5MLEA 0 CZK
SIAL479756-5ML
1-Methoxy-2-butanol, Sigma-Aldrich®
1-Methoxy-2-butanol
Vzorec: C₅H₁₂O₂
MW: 104,15 g/mol
MDL Number: MFCD00059143
CAS číslo: 53778-73-7
EINECS: 258-763-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR