Tisk…
Nonylphenol (mixture of isomers), PESTANAL® analytical standard, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
46018-1GEA 2270 CZK
46018-1G
Nonylphenol (mixture of isomers), PESTANAL® analytical standard, Supelco®
Nonylphenol (mixture of isomers)
Vzorec: C₁₅H₂₄O
MW: 220,35 g/mol
MDL Number: MFCD00002396
CAS číslo: 84852-15-3
EINECS: 284-325-5
UN: 3145
ADR: 8,II