Tisk…

Erucamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL280577-100GEA 0 CZK
SIAL280577-100G SIAL280577-5G
Erucamide, Sigma-Aldrich®
Erucamide
Vzorec: C₂₂H₄₃NO
MW: 337,5829 g/mol
MDL Number: MFCD00882379
CAS číslo: 112-84-5
EINECS: 204-009-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR