Tisk…

Mesitylacetic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL554286-25GEA 0 CZK
SIAL554286-25G
Mesitylacetic acid, Sigma-Aldrich®
Mesitylacetic acid
Vzorec: C₁₁H₁₄O₂
MW: 178,23
MDL Number: MFCD00014368
CAS číslo: 4408-60-0
EINECS: 224-556-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR