Tisk…

D-(+)-Cellobiose, Millipore®

Dodavatel: Merck

SIAL56481-10X25DISEA 0 CZK
SIAL56481-10X25DIS
D-(+)-Cellobiose, Millipore®
D-(+)-Cellobiose
Vzorec: C₁₂H₂₂O₁₁
MW: 342,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS číslo: 528-50-7
EINECS: 208-436-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR