Bismuth(III) isopropoxide
Dodavatel: Thermo Scientific
|
Varování
|
Packaged under argon.
43938.06EA
20520
CZK
43938.06
Bismuth(III) isopropoxide
Bismuth(III) isopropoxide
Vzorec:
C₉H₂₁BiO₃ MW: 386,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00799065 CAS číslo: 15049-67-9 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...