Tisk…

Decyl chloroformate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL549339-5GEA 0 CZK
SIAL549339-5G
Decyl chloroformate, Sigma-Aldrich®
Decyl chloroformate
Vzorec: C₁₁H₂₁ClO₂
MDL Number: MFCD00126853
CAS číslo: 55488-51-2
EINECS: 259-668-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR