Tisk…

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine, Sigma-Aldrich®

Dodavatel: Merck

SIAL74231-500MG-FEA 0 CZK
SIAL74231-500MG-F
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine, Sigma-Aldrich®
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Vzorec: C₂₄H₉F₁₈P
MW: 670,28 g/mol
MDL Number: MFCD00042428
CAS číslo: 175136-62-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR