4'-Ethoxy-2',3'-difluoroacetophenone 97%, JRD
|
Varování
|
H32495.06EA
7420
CZK
H32495.06
H32495.03
4'-Ethoxy-2',3'-difluoroacetophenone 97%, JRD
4'-Ethoxy-2',3'-difluoroacetophenone
Vzorec:
C₁₀H₁₀F₂O₂ MW: 200,18 g/mol Bod tání: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258710 CAS číslo: 1017779-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...