L(-)-Fucose
A4365.0100EA
63300
CZK
A4365.0100
L(-)-Fucose
L(-)-Fucose
Vzorec:
C₆H₁₂O₅ MW: 164,16 g/mol |
MDL Number:
MFCD00069812 CAS číslo: 2438-80-4 EINECS: 219-452-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...