Tisk…

4'-Methoxy-2-chloroacetophenone, Sigma-Aldrich®

Dodavatel: Merck

SIAL716618-1GEA 1960 CZK
SIAL716618-1G
4'-Methoxy-2-chloroacetophenone, Sigma-Aldrich®
4'-Methoxy-2-chloroacetophenone
Vzorec: C₉H₉ClO₂
MW: 184,6196 g/mol
MDL Number: MFCD00216508
CAS číslo: 2196-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR