Tisk…

5-Chloro-2-hydroxybenzophenone, Sigma-Aldrich®

Dodavatel: Merck

SIALC44702-5GEA 0 CZK
SIALC44702-5G
5-Chloro-2-hydroxybenzophenone, Sigma-Aldrich®
5-Chloro-2-hydroxybenzophenone
Vzorec: C₁₃H₉ClO₂
MW: 232,6600 g/mol
MDL Number: MFCD00020134
CAS číslo: 85-19-8
EINECS: 201-592-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR