Tisk…

4,4'-Diphenylbenzophenone ≥97%

Dodavatel: Thermo Scientific

L06502.06EA 1590 CZK
L06502.06
4,4'-Diphenylbenzophenone ≥97%
4,4'-Diphenylbenzophenone
Vzorec: C₂₅H₁₈O
Bod tání: 235…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00016575
CAS číslo: 3478-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR