Tisk…

6-Chloroquinolin-2(1H)-one, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL557773-5GEA 9390 CZK
SIAL557773-5G
6-Chloroquinolin-2(1H)-one, Sigma-Aldrich®
6-Chloroquinolin-2(1H)-one
Vzorec: C₉H₆ClNO
MW: 179,6030 g/mol
MDL Number: MFCD02930633
CAS číslo: 1810-67-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR