Tisk…

2-Chloro-5-fluoronicotinic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL697338-1GEA 3270 CZK
SIAL697338-1G
2-Chloro-5-fluoronicotinic acid, Sigma-Aldrich®
2-Chloro-5-fluoronicotinic acid
Vzorec: C₆H₃ClFNO₂
MW: 175,5449 g/mol
MDL Number: MFCD03092932
CAS číslo: 38186-88-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR