Tisk…

2-(p-Tolyl)pyridine, Sigma-Aldrich®

Dodavatel: Merck

SIAL198870-5GEA 1750 CZK
SIAL198870-5G SIAL198870-25G
2-(p-Tolyl)pyridine, Sigma-Aldrich®
2-(p-Tolyl)pyridine
Vzorec: C₁₂H₁₁N
MW: 169,22 g/mol
MDL Number: MFCD00006281
CAS číslo: 4467-06-5
EINECS: 224-734-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR