(R)-(+)-Pulegone
Dodavatel: LGC Standards PROMOCHEM
PROOMM0163.12EA
0
CZK
PROOMM0163.12
(R)-(+)-Pulegone
(R)-(+)-Pulegone
Vzorec:
C₁₀H₁₆O MW: 152,23 g/mol |
MDL Number:
MFCD00063000 CAS číslo: 89-82-7 EINECS: 201-943-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...