Tisk…

(Formylmethyl)triphenylphosphonium chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL47718-10GEA 1930 CZK
SIAL47718-10G
(Formylmethyl)triphenylphosphonium chloride, Sigma-Aldrich®
(Formylmethyl)triphenylphosphonium chloride
Vzorec: C₂₀H₁₈ClOP
MW: 340,79 g/mol
MDL Number: MFCD00012003
CAS číslo: 62942-43-2
EINECS: 263-767-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR