Tisk…

N-Hydroxyurethane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL119474-25GEA 0 CZK
SIAL119474-25G SIAL119474-5G
N-Hydroxyurethane, Sigma-Aldrich®
N-Hydroxyurethane
Vzorec: C₃H₇NO₃
MDL Number: MFCD00002108
CAS číslo: 589-41-3
EINECS: 209-648-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR