3-Piperidylpropiophenone hydrochloride
Dodavatel: LGC Standards PROMOCHEM
PROOBP450EA
7770
CZK
PROOBP450
3-Piperidylpropiophenone hydrochloride
3-Piperidylpropiophenone hydrochloride
Vzorec:
C₁₄H₂₀ClNO MW: 253,7677 g/mol |
MDL Number:
MFCD00035385 CAS číslo: 886-06-6 EINECS: 212-947-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...