Dodecacarbonyltriiron stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
Stabilization:
Stabilised with methanol
M-Clarity™ quality level = MQ100
SIAL381411-25GEA
0
CZK
SIAL381411-25G
SIAL381411-5G
Dodecacarbonyltriiron stabilizovaný, Sigma-Aldrich®
Dodecacarbonyltriiron
Vzorec:
C₁₂Fe₃O₁₂ MW: 503,66 g/mol |
MDL Number:
MFCD00135617 CAS číslo: 17685-52-8 EINECS: 241-668-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...