5-Fluoro-3-methylsalicylaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL593281-1GEA
7070
CZK
SIAL593281-1G
5-Fluoro-3-methylsalicylaldehyde, Sigma-Aldrich®
5-Fluoro-3-methylsalicylaldehyde
Vzorec:
C₈H₇FO₂ MW: 154,1384 g/mol |
MDL Number:
MFCD04039244 CAS číslo: 704884-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...