Tisk…

Hydrocortisone-21-acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIALH3035-5GEA 11760 CZK
SIALH3035-5G
Hydrocortisone-21-acetate, Sigma-Aldrich®
Hydrocortisone-21-acetate
Vzorec: C₂₃H₃₂O₆
MW: 404,51 g/mol
MDL Number: MFCD00037714
CAS číslo: 50-03-3
EINECS: 200-004-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR