1-Hydroxy-7-azabenzotriazole (HOAt)
MOLEM45186173EA
5950
CZK
MOLEM45186173
1-Hydroxy-7-azabenzotriazole (HOAt)
1-Hydroxy-7-azabenzotriazole (HOAt)
Vzorec:
C₅H₄N₄O MW: 136,1 g/mol |
MDL Number:
MFCD00210053 CAS číslo: 39968-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...