4-Nitrophenol-[2,3,5,6-D₄]
Dodavatel: EHRENSTORFER GMBH
|
Varování
|
EHERC15590404EA
6010
CZK
EHERC15590404
4-Nitrophenol-[2,3,5,6-D₄]
4-Nitrophenol-[2,3,5,6-D₄]
Vzorec:
C₆HD₄NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00143352 CAS číslo: 93951-79-2 UN: 1663 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...