Tisk…

Trihexyl(tetradecyl)phosphonium decanoate, Sigma-Aldrich®

Dodavatel: Merck

SIAL50826-50G-FEA 0 CZK
SIAL50826-50G-F
Trihexyl(tetradecyl)phosphonium decanoate, Sigma-Aldrich®
Trihexyl(tetradecyl)phosphonium decanoate
Vzorec: C₄₂H₈₇O₂P
MW: 655,113 g/mol
MDL Number: MFCD04117698
CAS číslo: 465527-65-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR