5-Amino-4-cyano-1-phenyl-1H-pyrazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL389595-1GEA
1130
CZK
SIAL389595-1G
5-Amino-4-cyano-1-phenyl-1H-pyrazole, Sigma-Aldrich®
5-Amino-4-cyano-1-phenyl-1H-pyrazole
Vzorec:
C₁₀H₈N₄ MW: 184,20 g/mol |
MDL Number:
MFCD00020730 CAS číslo: 5334-43-0 EINECS: 226-253-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...